Order Entry
Finland
ContactUsLinkComponent
 
 
 
Potassium hydroxide, pellets, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis (max. 0.002% Na)
 :  Merck
 
 
 
Potassium hydroxide, pellets, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis (max. 0.002% Na)
 :  Merck

 

  • Solubility:
    1130 g/l (20 °C)
  • Melting Point:
    360 °C
  • Molar Mass:
    56.11 g/mol
  • Boiling Point:
    1320 °C
  • Vapor Pressure:
    (20 °C)
  • Density:
    2.04 g/cm3 (20 °C)
  • pH:
    14 (56 g/l, H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material