



Hydrogen chloride 5 - 10% (w/w) in methanol for esterification
Supplier: TCI
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
:36.46 g/mol
Merck Index:14,04780
Storage Temperature:Refrigerator
EINECS:231-595-7
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (Neutralization titration):5.0 to 10.0 w/w%