Order Entry
Finland
ContactUsLinkComponent
2-Furaldehyde
2-Furaldehyde
Catalog # ROTH5080.1
Supplier:  Roth Carl
CAS Number:  
2-Furaldehyde
Catalog # ROTH5080.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

 :
 :96.09 g/mol
Merck Index:13,04325
Boiling Pt:162 °C (1013 hPa)
Melting Pt:–39 °C
Density:1.15 g/cm³ (20 °C)
Flash Pt:60 °C
Storage Temperature:Ambient
EINECS:202-627-7
UN:1199
ADR:6.1,II
 :GHS06
 :GHS08
CAS Number:98-01-1
Formula:C₅H₄O₂
MDL Number:MFCD00003229