Order Entry
Finland
ContactUsLinkComponent
Dibenzosuberone for synthesis
Dibenzosuberone for synthesis
Catalog # 8.03559.0050
Supplier:  Merck
CAS Number:  
Dibenzosuberone for synthesis
Catalog # 8.03559.0050
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Solubility:
    0.03 g/l (20 °C)
  • Melting Point:
    31 - 34 °C
  • Molar Mass:
    208.25 g/mol
  • Boiling Point:
    191 - 194 °C (7 hPa)
  • Flash Point:
    200 °C
  • Refractive Index:
    1.6324 (20 °C, 589 nm)
  • Density:
    1.16 g/cm3 (25 °C)

Specification Test Results

About this item

 :
 :208.26 g/mol
Boiling Pt:378 °C (1013 hPa)
Melting Pt:31…34 °C
Density:1.1635 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:214-912-3
CAS Number:1210-35-1
Formula:C₁₅H₁₂O
MDL Number:MFCD00003587