Order Entry
Finland
ContactUsLinkComponent
Dibenzosuberone 97%
Irritant
Dibenzosuberone 97%
Catalog # ACRO114741000
CAS Number:  
undefined
Irritant
Dibenzosuberone 97%
Catalog # ACRO114741000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Appearance:
    Clear yellow Liquid or low melting solid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥96.0 %
  • Titration with NaOH:
    ≥96.0 %

Specification Test Results

About this item

 :
 :208.26 g/mol
Boiling Pt:148 °C (.3 mmHg)
Melting Pt:32…34 °C
Density:1.15 g/cm³
Storage Temperature:Ambient
EINECS:214-912-3
 :GHS07
CAS Number:1210-35-1
Formula:C₁₅H₁₂O
MDL Number:MFCD00003587