
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222) ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:23978-09-8
:
Formula:C₁₈H₃₆N₂O₆
MDL Number:MFCD00005111
:376.49 g/mol
Melting Pt:69…71 °C
EINECS:245-962-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:69.0 to 73.0 deg-C