
4,4-Dimethyl-2-cyclohexen-1-one 97%
Catalog # ACRO319540050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4710 to 1.4750 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
About this item
:
:124.18 g/mol
Boiling Pt:72…73.5 °C (20 mmHg)
Density:0.94 g/cm³
Flash Pt:64 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1073-13-8
Formula:C₈H₁₂O
MDL Number:MFCD00009695