Order Entry
Finland
ContactUsLinkComponent
 
 
3-Methyl-2-cyclohexen-1-one 98%
 
undefined
 
3-Methyl-2-cyclohexen-1-one 98%

 

  • Appearance:
    Clear colorless to light yellow to brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4935 to 1.4955 (20°C, 589 nm)
  • GC:
    ≥97.5 %