Order Entry
Finland
ContactUsLinkComponent
 
 
 
2-Cyclohexen-1-one 97%
 
undefined
 
 
2-Cyclohexen-1-one 97%

 

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4875 to 1.4895 (20°C, 589 nm)
  • GC:
    ≥96.0 %
  • Absorption Ratio P-15/P+15:
    1.01 to 1.16
  • BSC Certification:
    Passes