(1S)-(-)-α-Pinene ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:7785-26-4
:
Formula:C₁₀H₁₆
MDL Number:MFCD00064145
:136.24 g/mol
Merck Index:14,07445
Boiling Pt:155°C
Melting Pt:–62 °C
Density:0,8592 g/cm³ (20 °C)
Flash Pt:32°C
Storage Temperature:Refrigerator
EINECS:232-077-3
UN:2368
ADR:3,III
:GHS02
:GHS07
:GHS09
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
- Specific rotation [a]20/D:-39.0 to -52.0 deg (neat)