0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
2,6-Dimethylphenylboronic acid
2,6-Dimethylphenylboronic acid ≥97%
2,6-Dimethylphenylboronic acid ≥97%
:
Thermo Fisher Scientific
2,6-Dimethylphenylboronic acid ≥97%
:
Thermo Fisher Scientific
:
100379-00-8
:
:
C₈H₁₁BO₂
:
MFCD01009693
:
149.98 g/mol
:
120…126 °C (dec.)
:
GHS07
:
Ambient
msds_ALFAB24613
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years