Bromocresol purple
Catalog # CHRO3L-103
Supplier: CHROMA
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:540.23 g/mol
Merck Index:13,01371
Melting Pt:242 °C
EINECS:204-087-8
:GHS07
Storage Temperature:Ambient
CAS Number:115-40-2
Formula:C₂₁H₁₆Br₂O₅S
MDL Number:MFCD00011681