
4-Nitro-3-(trifluoromethyl)aniline
Catalog # MOLEM15949255
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:206.12 g/mol
Melting Pt:129 °C
Density:1.752 g/cm³ (–123.15 °C)
EINECS:206-884-6
UN:2811
ADR:6.1,III
:GHS07
CAS Number:393-11-3
Formula:C₇H₅F₃N₂O₂
MDL Number:MFCD00014717