Order Entry
Finland
Orders LinkContactUsLinkComponent
4'-Fluoroacetophenone

4'-Fluoroacetophenone

Glass bottle for solids, 100 g
Catalog # MOLEM58089548
Irritant
Marketsource
Supplier:   Molekula
4'-Fluoroacetophenone
Catalog # MOLEM58089548
Supplier:  Molekula
Marketsource
CAS Number:  403-42-9
4'-Fluoroacetophenone

Glass bottle for solids, 100 g

CAS Number:  403-42-9
Price
77,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1-(4-Fluorophenyl)ethanone, p-Fluoroacetophenone
  • CAS Number:403-42-9
  • Formula:C₈H₇FO
  • MDL Number:MFCD00000354
  • Molecular Weight:138.14 g/mol
  • Boiling Pt:196 °C (1013 hPa)
  • Density:1,138 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:206-960-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.