4'-Fluoroacetophenone 99%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 1-(4-Fluorophenyl)ethanone, p-Fluoroacetophenone
- CAS Number:403-42-9
- Formula:C₈H₇FO
- MDL Number:MFCD00000354
- Molecular Weight:138.14 g/mol
- Boiling Pt:196 °C (760 mmHg)
- Melting Pt:4 °C
- Density:1.13 g/cm³
- Flash Pt:71 °C
- Storage Temperature:Ambient
- EINECS:206-960-9
3 Options Available Below
Product Details & Documents
Synonyms: 1-(4-Fluorophenyl)ethanone, p-Fluoroacetophenone
- CAS Number:403-42-9
- Formula:C₈H₇FO
- MDL Number:MFCD00000354
- Molecular Weight:138.14 g/mol
- Boiling Pt:196 °C (760 mmHg)
- Melting Pt:4 °C
- Density:1.13 g/cm³
- Flash Pt:71 °C
- Storage Temperature:Ambient
- EINECS:206-960-9
Documents
ACRO11926
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5090 to 1.5120 (20°C, 589 nm)
- GC:≥98.5 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



