Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2'-Fluoroacetophenone
2'-Fluoroacetophenone ≥97%
2'-Fluoroacetophenone ≥97%
:
Thermo Fisher Scientific
2'-Fluoroacetophenone ≥97%
:
Thermo Fisher Scientific
:
445-27-2
:
:
C₈H₇FO
:
MFCD00000320
:
138.14 g/mol
:
82…83 °C (10 mmHg)
:
1.137
:
61 °C (141 °F)
:
207-156-0
:
GHS07
:
Ambient
msds_ALFAA15128
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years