2,5-Dichlorobenzyl alcohol ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:34145-05-6
:
Formula:C₇H₆Cl₂O
MDL Number:MFCD00004607
:177.03 g/mol
Boiling Pt:264 °C (1013 hPa)
Melting Pt:76…80 °C
Storage Temperature:Ambient
EINECS:251-850-6
:GHS07