Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
α-Bromo-4-iodotoulene
α-Bromo-4-iodotoulene
α-Bromo-4-iodotoulene
α-Bromo-4-iodotoulene
MOLE22950982-5G
:
Molekula
:
α-Bromo-4-iodotoulene
MOLE22950982-5G
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
296.93 g/mol
:
115…120 °C (0.6 mmHg)
:
80 °C
:
Refrigerator
:
3261
:
8,II
:
GHS05
:
16004-15-2
:
C₇H₆BrI
:
MFCD00209656