
α-Bromo-4-iodotoulene 97%
:16004-15-2
:
:C₇H₆BrI
:MFCD00209656
:296.93 g/mol
:140 °C (15 mmHg)
:77…82 °C
:Refrigerator
:3261
:8,II
:GHS05
- Appearance:White to light yellow Powder or crystals
- Infrared spectrum:Conforms
- Melting point:77 to 82 °C
- GC:≥96.0 %