Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
ω-Bromo-4-methoxyacetophenone
ω-Bromo-4-methoxyacetophenone ≥98%
ω-Bromo-4-methoxyacetophenone ≥98%
:
Thermo Fisher Scientific
ω-Bromo-4-methoxyacetophenone ≥98%
:
Thermo Fisher Scientific
:
2632-13-5
:
:
C₉H₉BrO₂
:
MFCD00000201
:
229.07 g/mol
:
285 °C (1013 hPa)
:
68…72 °C
:
Refrigerator
:
220-118-8
:
3261
:
8,II
:
GHS05
msds_ALFAA13415
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years