

ω-Bromo-4-methoxyacetophenone 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2632-13-5
:
Formula:C₉H₉BrO₂
MDL Number:MFCD00000201
:229.07 g/mol
Boiling Pt:285 °C (1013 hPa)
Melting Pt:69…72 °C
Storage Temperature:Refrigerator
EINECS:220-118-8
UN:3261
ADR:8,II
:GHS07
:GHS05
Specification Test Results
- Appearance:Off-white to light brown Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:69 to 72 °C
- GC:≥97.5 %