
ω-Bromo-3-methoxyacetophenone ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5000-65-7
:
Formula:C₉H₉BrO₂
MDL Number:MFCD00000199
:229.07 g/mol
Boiling Pt:285 °C (1013 hPa)
Melting Pt:60…64 °C
Storage Temperature:Refrigerator
EINECS:225-666-1
UN:3261
ADR:8,II
:GHS05