Order Entry
Finland
ContactUsLinkComponent
 
 
 
ω-Bromoacetophenone 98%
 
undefined
 
 
ω-Bromoacetophenone 98%

 

  • Appearance:
    White to green-brown Powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    46 to 51 °C
  • GC:
    ≥97.5 %