
N,N,N',N'-Tetramethyl-p-phenylenediamine 98%
:100-22-1
:
:C₁₀H₁₆N₂
:MFCD00008309
:164.25 g/mol
:12,09367
:260 °C (760 mmHg)
:48…52 °C
:1,08 g/cm³ (20 °C)
:>112 °C
:Refrigerator
:202-831-6
:2811
:6.1,III
:GHS07
- Appearance:Beige to grey to purple to brown Crystals or flakes or granular solid
- Infrared spectrum:Conforms
- Melting point:48 to 52 °C
- GC:≥97.5 %
Frequently Bought Together