

3-Iodo-4-nitroanisole 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:214279-40-0
:
Formula:C₇H₆INO₃
MDL Number:MFCD01320682
:279.03 g/mol
Melting Pt:68…73 °C
UN:2811
ADR:6.1,III
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow to orange-brown Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:68 to 73 °C
- GC:≥96.0 %