2-Fluorophenyl isocyanate
Catalog # MAYBSB01843DA
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Boiling Pt:41…44 °C (1 mmHg)
Density:1.222 g/cm³ (20 °C)
Storage Temperature:Refrigerator
UN:3080
ADR:6.1,II
:137.11 g/mol
EINECS:240-804-0
CAS Number:16744-98-2
Formula:C₇H₄FNO
MDL Number:MFCD00001996