2-Fluorophenyl isocyanate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16744-98-2
:
Formula:C₇H₄FNO
MDL Number:MFCD00001996
Boiling Pt:56…58 °C (10 mmHg)
Density:1.221
Flash Pt:50 °C (122 °F)
Storage Temperature:Refrigerator
EINECS:240-804-0
UN:3080
ADR:6.1,II
:137.11 g/mol
:GHS06
:GHS08
:GHS02
:GHS07