Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Kirjaudu sisään
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-((Trifluoromethyl)thio)benzoic acid
3-((Trifluoromethyl)thio)benzoic acid ≥97%
3-((Trifluoromethyl)thio)benzoic acid ≥97%
Irritant
3-((Trifluoromethyl)thio)benzoic acid ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
3-((Trifluoromethyl)thio)benzoic acid ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
946-65-6
:
Formula:
C₈H₅F₃O₂S
MDL Number:
MFCD00236337
:
222.19 g/mol
Melting Pt:
71…74 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAA19975
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation