Bottles, wide neck, with screw cap, round
Supplier: RIXIUS
Certificates
About this item
Clear and amber glass with thread and black PP cap.
- Good chemical and physical properties
- For applications that do not involve high thermal stresses
- Round thread in accordance with DIN 168
Ideal for sampling, storing and transporting powders and pastes.