Order Entry
Finland
ContactUsLinkComponent
Dimethyl terephthalate for synthesis, Sigma-Aldrich®
Dimethyl terephthalate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Dimethyl terephthalate for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    0.036 g/l (20 °C)
  • Melting Point:
    139 - 141 °C
  • Molar Mass:
    194.19 g/mol
  • Bulk Density:
    500 kg/m3
  • Boiling Point:
    282 °C (1013 hPa)
  • Vapor Pressure:
     - 1 hPa (20 °C)
  • Flash Point:
    154 °C (External MSDS)
  • Explosion Limit:
    0.8 - 11.8 %(V)
  • Density:
    1.35 g/cm3 (20 °C)
  • Ignition Point:
    520 °C DIN 51794

Specification Test Results

Related Information