Order Entry
Finland
ContactUsLinkComponent
 
Benzanilide
Catalog # PROOICRS0221
CAS Number:  
Benzanilide
Catalog # PROOICRS0221
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    4 g
  • Seq:
    3155

Specifications

About this item

 :
Merck Index:13,01060
Boiling Pt:247 °C (1013 hPa)
Melting Pt:162…165 °C
Density:1.32 g/cm³ (20 °C)
 :197.24 g/mol
EINECS:202-292-7
CAS Number:93-98-1
Formula:C₁₃H₁₁NO
MDL Number:MFCD00003069