Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Cyanobenzenesulphonyl chloride
4-Cyanobenzenesulphonyl chloride
4-Cyanobenzenesulphonyl chloride
4-Cyanobenzenesulphonyl chloride
Supplier:
Thermo Scientific
Certificates
Expand image
4-Cyanobenzenesulphonyl chloride
Supplier:
Thermo Scientific
About this item
CAS Number:
49584-26-1
:
Formula:
C₇H₄ClNO₂S
MDL Number:
MFCD00052337
:
201.63 g/mol
UN:
3261
ADR:
8,II