
4-Chlororesorcinol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:95-88-5
:
Formula:C₆H₅ClO₂
MDL Number:MFCD00002273
:144.56 g/mol
Boiling Pt:147 °C (18 mmHg)
Melting Pt:106…110 °C
Flash Pt:148 °C
Storage Temperature:Ambient
EINECS:202-462-0
:GHS07
Specification Test Results
- Appearance:Beige Powder
- Infrared spectrum:Conforms
- Melting point:106 to 110 °C
- GC:≥97.5 %