Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Dichlorophenyl isothiocyanate
2,5-Dichlorophenyl isothiocyanate
2,5-Dichlorophenyl isothiocyanate
MAYBSB01698EA
:
Thermo Scientific
:
2,5-Dichlorophenyl isothiocyanate
MAYBSB01698EA
:
Thermo Scientific
:
Pack type:
Glass bottle for solids
Pk:
10 g
Seq:
3236
:
:
204.08 g/mol
:
145…147 °C (15 mmHg)
:
1.425 g/cm³ (20 °C)
:
3334
:
9,
:
GHS05
:
3386-42-3
:
C₇H₃Cl₂NS
:
MFCD00041054