

2,4-Dichlorophenyl isothiocyanate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6590-96-1
:
Formula:C₇H₃Cl₂NS
MDL Number:MFCD00037827
Boiling Pt:130…133 °C (2 mmHg)
Melting Pt:36…39 °C
Density:1.410
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:229-527-6
ADR:8,III
:204.08 g/mol
UN:2923
:GHS06
:GHS05