

2,6-Dichlorophenyl isothiocyanate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6590-95-0
:
Formula:C₇H₃Cl₂NS
MDL Number:MFCD00041056
:204.08 g/mol
Boiling Pt:122…124 °C (8 mmHg)
Melting Pt:42…44 °C
Storage Temperature:Refrigerator
UN:2923
ADR:8,III
:GHS06
:GHS05