
2,3-Dichlorophenyl isothiocyanate
Catalog # MAYBSB01755EA
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
Boiling Pt:256…258 °C (1013 hPa)
Density:1.434 g/cm³ (20 °C)
ADR:8,III
:204.08 g/mol
UN:3265
:GHS05
CAS Number:6590-97-2
Formula:C₇H₃Cl₂NS
MDL Number:MFCD00041048