

2,3-Dichlorophenyl isothiocyanate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6590-97-2
:
Formula:C₇H₃Cl₂NS
MDL Number:MFCD00041048
Boiling Pt:256…258 °C
Density:1.434
Flash Pt:>110 °C (230 °F)
UN:2922
ADR:8,III
:204.08 g/mol
:GHS06
:GHS05
Storage Temperature:Ambient