
4-(Methylthio)benzaldehyde 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3446-89-7
:
Formula:C₈H₈OS
MDL Number:MFCD00006948
:152.22 g/mol
Boiling Pt:86…90 °C (1 mmHg)
Melting Pt:6 °C
Density:1.14 g/cm³
Flash Pt:>110 °C
EINECS:222-365-7
ADR:9,
:GHS07
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.6440 to 1.6500 (20°C, 589 nm)
- GC:≥96.0 %