Order Entry
Finland
ContactUsLinkComponent
2,4-Dibromo-6-fluoroaniline
2,4-Dibromo-6-fluoroaniline
Catalog # MAYBBTB06640EA
Supplier:  Thermo Scientific
CAS Number:  
2,4-Dibromo-6-fluoroaniline
Catalog # MAYBBTB06640EA
Supplier:  Thermo Scientific
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
Boiling Pt:277 °C (1013 hPa)
Melting Pt:62…64 °C
ADR:6.1,III
 :268.91 g/mol
UN:2811
CAS Number:141474-37-5
Formula:C₆H₄Br₂FN
MDL Number:MFCD00042230