Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-4-nitrobenzoic acid
3-Bromo-4-nitrobenzoic acid ≥97%
3-Bromo-4-nitrobenzoic acid ≥97%
Irritant
3-Bromo-4-nitrobenzoic acid ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
3-Bromo-4-nitrobenzoic acid ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
101420-81-9
:
Formula:
C₇H₄BrNO₄
MDL Number:
MFCD04117949
:
246.02 g/mol
Melting Pt:
200…204 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH27268
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation