Order Entry
Finland
ContactUsLinkComponent
1-Ethyl-2-pyrrolidone 98%
Corrosive
Health
1-Ethyl-2-pyrrolidone 98%
Certificates
undefined
Corrosive
Health
1-Ethyl-2-pyrrolidone 98%

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4640 to 1.4680 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • HPLC:
    ≥97.5 %

Specification Test Results

Related Information