
2-Pyrrolidone for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:616-45-5
:
Formula:C₄H₇NO
MDL Number:MFCD00005270
:85.11 g/mol
Merck Index:12,08200
Boiling Pt:245 °C (1013 hPa)
Melting Pt:24…26 °C
Density:1.1115 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:129 °C
EINECS:210-483-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:24 - 26 °C
- Molar Mass:85.11 g/mol
- Boiling Point:250 °C (1013 hPa)
- Vapor Pressure:0.04 hPa (20 °C)
- Flash Point:138 °C
- Refractive Index:1.48 (20 °C, 589 nm)
- Explosion Limit:1.8 - 16.6 %(V)
- Density:1.11 g/cm3 (20 °C)
- pH:9 - 11 (100 g/l, H2O, 20 °C)
- Ignition Point:390 °C