9-Nitroanthracene
Glass bottle, 1,2 mL
Catalog # PROOCILDLM47121.2
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
- CAS Number:602-60-8
- Formula:C₁₄H₉NO₂
- MDL Number:MFCD00001248
- Molecular Weight:223.23 g/mol
- Boiling Pt:403 °C (1013 hPa)
- Melting Pt:137…139 °C
- Density:1.591 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:210-021-9
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL



