
3,4,5-Trimethoxycinnamic acid predominantly trans 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:90-50-6
:
Formula:C₁₂H₁₄O₅
MDL Number:MFCD00004388
:238.24 g/mol
Boiling Pt:367 °C (1013 hPa)
Melting Pt:126…128 °C
Storage Temperature:Ambient
EINECS:201-999-8
:GHS07
Related Information
Frequently Bought Together