
3,4-Dimethoxycinnamic acid predominantly trans 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2316-26-9
:
Formula:C₁₁H₁₂O₄
MDL Number:MFCD00004387
:208.21 g/mol
Boiling Pt:343 °C (1013 hPa)
Melting Pt:181…184 °C
EINECS:219-025-5
:GHS07
Specification Test Results
- Appearance:Light yellow to light beige Powder
- Infrared spectrum:Conforms
- Melting point:181 to 184 °C
- GC:≥98.5 %
- Titration with NaOH:≥98.5 %