

4-Methyl-1-pentene ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:691-37-2
:
Formula:C₆H₁₂
MDL Number:MFCD00008949
:84.16 g/mol
Boiling Pt:53°C
Melting Pt:–153.6 °C
Density:0,665 g/cm³ (20 °C)
Flash Pt:–32°C
Storage Temperature:Ambient
EINECS:211-720-1
UN:2288
ADR:3,II
:GHS02
:GHS08
Specification Test Results
- Purity (GC):min. 97.0 %
- Appearance:Colorless clear liquid