Order Entry
Finland
ContactUsLinkComponent
Bisacrylamide 96%, extra pure
Irritant
Bisacrylamide 96%, extra pure
Certificates
undefined
Irritant
Bisacrylamide 96%, extra pure
Certificates

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤1 % (K.F.)
  • HPLC:
    ≥95.0 %

Specification Test Results

Related Information