

Acrylamide
Supplier: IBI SCIENTIFIC
Certificates
About this item
CAS Number:79-06-1
:
Formula:C₃H₅NO
MDL Number:MFCD00008032
:71.08 g/mol
Boiling Pt:232 °C (1013 hPa)
Melting Pt:84.5 °C
Density:1.122 g/cm³ (20 °C)
Flash Pt:138 °C
Storage Temperature:Refrigerator
EINECS:201-173-7
UN:2074
ADR:6.1,III
:GHS06
:GHS08