Mounting medium, kaiser's glycerol gelatine, phenol-free
Catalog # 1.08635.0100
Supplier: Merck
CAS Number:
Specifications
- Pk:100 g
- Description:Kaiser's glycerol gelatine, phenol-free mounting medium
Specifications
About this item
Kaiser's glycerol gelatin is an aqueous mounting medium for microscopy and works well with specimens prepared for enzyme, lipid, and immunohistochemistry determinations.
- Density: 1,117 to 1,120 g/cm³ (40 °C)
- Storage: +15 °C to +25 °C
The preservative phenol is replaced with a proprietary compound that is more environmentally friendly. This affords the unique advantage of eliminating phenol fumes when the medium is heated before use. It is developed specifically for use in histological applications is essential in the support of formal quality assurance programs to monitor staining performance in pathology laboratories.
Preparation Capacity: 400 slides.